Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentanecarbonyl chloride, 98%

Catalog No. AAA1414214
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1414214 25 g
AAA1414206 5 g
AAA1414222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1414214 Supplier Thermo Scientific Chemicals Supplier No. A1414214
Add to Cart

CAS: 4524-93-0 | C6H9ClO | 132.587 g/mol

Cyclopentanecarbonyl chloride is used in biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentanecarbonyl chloride is used in biochemical for proteomics research.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store in cool, dry place in tightly closed containers. Store away from oxidizing agent and water/moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4524-93-0
Molecular Formula C6H9ClO
Molecular Weight (g/mol) 132.587
MDL Number MFCD00001370
InChI Key WEPUZBYKXNKSDH-UHFFFAOYSA-N
Synonym cyclopentanecarbonylchloride, cyclopentane carboxyl chloride, cyclopentanoyl chloride, pubchem13237, acmc-1csoo, cyclopentylformyl chloride, cyclopetanecarbonylchloride, cyclopentancarbonyl chloride, cyclopentylcarbonyl chloride, chlorocarbonyl cyclopentane
PubChem CID 78284
IUPAC Name cyclopentanecarbonyl chloride
SMILES C1CCC(C1)C(=O)Cl

Specifications

Density 1.091
Boiling Point 160°C to 162°C
Flash Point 60°C (140°F)
Odor Acrid
Refractive Index 1.464
Quantity 25 g
UN Number UN2920
Beilstein 1237147
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 132.59
Percent Purity 98%
Chemical Name or Material Cyclopentanecarbonyl chloride
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber 224-856-1
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.