Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentanethiol 98.0+%, TCI America™
SDP

Catalog No. C081025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C081025ML 25 mL
C08105ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C081025ML Supplier TCI America Supplier No. C081025ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1679-07-8
Molecular Formula C5H10S
Molecular Weight (g/mol) 102.20
MDL Number MFCD00001369
InChI Key WVDYBOADDMMFIY-UHFFFAOYSA-N
Synonym cyclopentyl mercaptan, mercaptocyclopentane, cyclopentylthiol, fema no. 3262, cyclopentyl thiol, cyclopen-tanethiol, cyclopentane thiol, cyclopentylmercaptan, acmc-209dxb, cyclopentanethiol
PubChem CID 15510
IUPAC Name cyclopentanethiol
SMILES SC1CCCC1

Specifications

Color Colorless
Boiling Point 132°C
Quantity 25 mL
UN Number 1993
Formula Weight 102.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclopentanethiol
RTECSNumber GY4701000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.