Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentanone-2,2,5,5-d{4}, 95%

Catalog No. AAH5109503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5109503 1 g
AAH5109506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5109503 Supplier Thermo Scientific Chemicals Supplier No. H5109503
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 3997-89-5 | C5H8O | 88.142 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3997-89-5
Molecular Formula C5H8O
Molecular Weight (g/mol) 88.142
MDL Number MFCD00190415
InChI Key BGTOWKSIORTVQH-KHORGVISSA-N
Synonym cyclopentanone-2,2,5,5-d4, cyclopentanone d4, 2,2,5,5-∼2∼h_4_ cyclopentanone, cyclopentanone-2,2,5,5-d4, 98 atom % d, 2,2,5,5-2 h? cyclopentan-1-one
PubChem CID 16213326
IUPAC Name 2,2,5,5-tetradeuteriocyclopentan-1-one
SMILES C1CCC(=O)C1

Specifications

Quantity 1 g
UN Number UN2245
Formula Weight 88.14
Percent Purity 95%
Chemical Name or Material Cyclopentanone-2,2,5,5-d4
Hazard Category H226-H315-H319
Hazard Statement GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CYCLOPENTANONE
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.