Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentyl isocyanate, 97%

Catalog No. AC397370010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC397370010 1 g
AC397370050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC397370010 Supplier Thermo Scientific Chemicals Supplier No. 397370010
Add to Cart

CAS: 4747-71-1 | C6H9NO | 111.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4747-71-1
Molecular Formula C6H9NO
Molecular Weight (g/mol) 111.14
MDL Number MFCD02093695
InChI Key CZALJDQHONFVFU-UHFFFAOYSA-N
Synonym cyclopentyl isocyanate, cyclopentylisocyanate, cyclopentane, isocyanato, isocyanic acid cyclopentyl ester, isocyanato cyclopentane, isocyanato-cyclopentane, acmc-1aspa, ksc497s6f
PubChem CID 643500
IUPAC Name isocyanatocyclopentane
SMILES C1CCC(C1)N=C=O

Specifications

Density 0.9860g/mL
Boiling Point 147.0°C
Flash Point 36°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C5H9NCO
Quantity 1 g
Specific Gravity 0.986
Formula Weight 111.14
Percent Purity 97%
Chemical Name or Material Cyclopentyl isocyanate
Show More Show Less
Hazard Category GHS Signal Word: Danger

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.