Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentyl methyl ether, 99.5%, Extra Dry, stabilized, AcroSeal™

Catalog No. p-3757380
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC397251000 100 mL AcroSeal™ Glass Bottle
AC397250010 1 L AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC397251000 Supplier Thermo Scientific Chemicals Supplier No. 397251000
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 5614-37-9 | C6H12O | 100.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5614-37-9
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD08276401
InChI Key SKTCDJAMAYNROS-UHFFFAOYSA-N
Synonym cyclopentyl methyl ether, cyclopentane, methoxy, cpme, cyclopentane,methoxy, unii-4067e5gbkb, cyclopentylmethylether, cylopentylmethylether, methoxycyclopentane, 2-methoxycyclopentyl, cylopentyl-methylether
PubChem CID 138539
IUPAC Name methoxycyclopentane
SMILES COC1CCCC1

Specifications

Melting Point -140.0°C
Density 0.8600g/mL
Boiling Point 106.0°C
Flash Point −1°C
Assay Percent Range 99.4% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula C5H9OCH3
Quantity 100 mL
UN Number 1993
Residue after Evaporation 0.005% max.
Specific Gravity 0.86
Merck Index 15, 2742
Stabilizer 50ppm (BHT) approximately
Formula Weight 100.16
Percent Purity 99.5%
Grade Extra Dry
Physical Form Liquid
Chemical Name or Material Cyclopentyl methyl ether, Stabilized, Extra Dry
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May form explosive peroxides.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye protection/face p

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.