Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentylacetic Acid 98.0+%, TCI America™
SDP

Catalog No. C089425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C089425G 25 g
C08945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C089425G Supplier TCI America Supplier No. C089425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1123-00-8
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00001387
InChI Key YVHAIVPPUIZFBA-UHFFFAOYSA-N
Synonym cyclopentylacetic acid, cyclopentaneacetic acid, cycylopentylacetic acid, carboxymethyl cyclopentane, cyclopentyl acetic acid, cyclopentyacetic acid, cyclopentanacetic acid, cyclopentyl-acetic acid, 2-cyclopentylaceticacid, cyclopentane acetic acid
PubChem CID 71606
IUPAC Name 2-cyclopentylacetic acid
SMILES OC(=O)CC1CCCC1

Specifications

Color Yellow
Boiling Point 231°C
Quantity 25 g
Formula Weight 128.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Cyclopentylacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.