Learn More
Description
PA1-028 detects cyclophilin D (CyPD) in rat, hamster, canine, mouse, and human tissues. PA1-028 has been sucessfully used in Western blot procedures. By Western blot, this antibody detects a ~15 kDa protein in rat liver and heart extract, and a ~37 kDa in rat brain extract. The PA1-028 immunogen is a synthetic peptide corresponding to residues S(42) G N P L V Y L D V D A N(54) C of mature human CyPD. This peptide (Cat. # PEP-104) is available for use in neutralization and control experiments.
Specifications
Specifications
| Antigen | Cyclophilin 40 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide |
| Gene | PPID |
| Gene Accession No. | Q08752, Q6DGG0, Q9CR16 |
| Gene Alias | 40 kDa peptidyl-prolyl cis-trans isomerase; 40 kDa peptidyl-prolyl cis-trans isomerase D; 4930564J03Rik; cyclophilin 40; cyclophilin D; cyclophilin-40; Cyclophilin-related protein; Cyp 40; Cyp D; CYP40; CYP-40; CYPD; CYP-S1; cytoplasmic cyclophilin D; EC 5.2.1.8; I79_011990; MGC33096; peptidyl-prolyl cis-trans isomerase D; peptidylprolyl isomerase D; peptidylprolyl isomerase D (cyclophilin D); PPIase; PPIase D; PPID; Ppidl; Ppif; rotamase; Rotamase D; S-cyclophilin; SCYLP; testicular tissue protein Li 147 |
| Gene Symbols | PPID |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
