Learn More
Enzo Life Sciences Cyclopiazonic acid (10mg). CAS: 18172-33-3
SDP
Supplier:  Enzo Life Sciences BMLCA4150010
Encompass_Preferred
Mycotoxin. Cell permeable. Induces the release of intracellular stored Ca2+ without increasing IP3 levels via inhibition of endoplasmic reticulum Ca2+-ATPase. Highly specific inhibitor of Ca2+-ATPase of sarcoplasmic reticulum, completely inhibiting the enzyme at 6-8 nmol/mg protein (at 0.5-2.0µM ATP). Alternative name: CPA. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (20mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0393
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.