Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopropanecarboxamide 98.0+%, TCI America™
SDP

Catalog No. C2010250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2010250G 250 g
C201025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2010250G Supplier TCI America Supplier No. C2010250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6228-73-5
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.106
InChI Key AIMMVWOEOZMVMS-UHFFFAOYSA-N
Synonym cyclopropylcarboxamide, carbamoylcyclopropane, cyclopropyl carboxamide, seed one, cyclopropanecarboxylic acid amide, cyclopropanamide, cyclopropancarboxamide, carbamoyl cyclopropane, cyclopropane carboxamide, pubchem21401
PubChem CID 80376
ChEBI CHEBI:51457
IUPAC Name cyclopropanecarboxamide
SMILES C1CC1C(=O)N

Specifications

Melting Point 122°C
Quantity 250 g
Formula Weight 85.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Cyclopropanecarboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.