Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopropyl Cyanide 97.0+%, TCI America™
SDP

Catalog No. C099725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C099725ML 25 mL
C09975ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C099725ML Supplier TCI America Supplier No. C099725ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5500-21-0
Molecular Formula C4H5N
Molecular Weight (g/mol) 67.091
MDL Number MFCD00001269
InChI Key AUQDITHEDVOTCU-UHFFFAOYSA-N
Synonym cyclopropyl cyanide, cyanocyclopropane, cyclopropylnitrile, cyclopropanenitrile, cyclopropane cyanide, 1-cyanocyclopropane, cyclopropane carbonitrile, cyclopropylcyanide, 1-cyanocyclopropyl, cyclopropancarbonitril
PubChem CID 79637
IUPAC Name cyclopropanecarbonitrile
SMILES C1CC1C#N

Specifications

Boiling Point 135°C
Quantity 25 mL
UN Number 3275
Formula Weight 67.09
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclopropyl Cyanide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.