Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopropylacetylene 98.0+%, TCI America™
SDP

Catalog No. C198425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C198425G 25 g
C19845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C198425G Supplier TCI America Supplier No. C198425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6746-94-7
Molecular Formula C5H6
Molecular Weight (g/mol) 66.103
MDL Number MFCD02181090
InChI Key NPTDXPDGUHAFKC-UHFFFAOYSA-N
Synonym cyclopropylacetylene, cyclopropyl acetylene, ethynyl cyclopropane, cyclopropane,ethynyl, cyclopropane, ethynyl, cyclopropylethyne, cyclopropyacetylene, ethynyl-cyclopropane, ethynylcyclo-propane, cyclopropylacethylene
PubChem CID 138823
IUPAC Name ethynylcyclopropane
SMILES C#CC1CC1

Specifications

Color Colorless
Boiling Point 52°C
Quantity 25 g
UN Number 1993
Formula Weight 66.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclopropylacetylene
EINECSNumber (3)-4334
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.