Learn More
Sigma Organic Chemistry cyclopropylcarbonyl ami | 10mg | 64513-70-8 | MFCD01724840
Supplier: Sigma Organic Chemistry CDS01333610MG

cyclopropylcarbonyl ami , 10mg
About this Item:
CAS #: 64513-70-8
MDL #: MFCD01724840
Molecular Weight: 143.14
UNSPSC Code: 12352200
Chemical Formula: C6H9NO3
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.