Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopropylmagnesium bromide, 0.5M solution in THF, AcroSeal™

Catalog No. AC397611000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC397611000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC397611000 Supplier Thermo Scientific Chemicals Supplier No. 397611000
  • $326.50
    $277.65 / Each of 1

    Save $48.85

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 23719-80-4 | C3H5BrMg | 145.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23719-80-4 , 109-99-9
Molecular Formula C3H5BrMg
Molecular Weight (g/mol) 145.28
MDL Number MFCD00672094
InChI Key QUPLLJGAJVXXBW-UHFFFAOYSA-M
Synonym cyclopropylmagnesium bromide, cyclopropylmagnesiumbromide, bromo cyclopropyl magnesium, magnesium, bromocyclopropyl, cyclopropyl magnesium bromide, cprmgbr, cyclopropyl magnesiumbromide, magnesium,bromocyclopropyl, bromo-cyclopropyl magnesium, cyclopropylmagnesiunn bromide, grignard reagent
PubChem CID 11116189
IUPAC Name magnesium;cyclopropane;bromide
SMILES Br[Mg]C1CC1

Specifications

Color Colorless or Yellow
Physical Form Solution
Quantity 100 mL
Linear Formula C3H5MgBr
Flash Point −17°C
Formula Weight 145.28
Concentration or Composition (by Analyte or Components) 0.4 to 0.6M
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.968
Density 0.9680g/mL
Chemical Name or Material Cyclopropylmagnesium bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spon
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.