Learn More
Apexbio Technology LLC Nalidixic acid 389-08-2 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B19855.1
Encompass_Preferred
Nalidixic acid is a quinolone-class antibacterial agent targeting bacterial DNA gyrase and topoisomerase IV enzymes essential for bacterial DNA replication and topology maintenance Its mechanism involves disruption of DNA supercoiling and replication processes exhibiting bacteriostatic activity at lower concentrations and bactericidal effects when concentration increases Nalidixic acid is typically employed in microbiological studies for antibacterial mechanistic research antibiotic resistance characterization and screening bacterial strains with altered gyrase or topoisomerase IV susceptibilities Reported IC50 values against DNA gyrase typically range from 10 to 50 M depending on bacterial species and experimental conditions
Catalog No. 50-000-62353
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.