Learn More
Selleck Chemical LLC Cyclosporine 10mM/1mL 79217-60-0
SDP
Supplier:  Selleck Chemical LLC S151410MM/1ML
Encompass_Preferred
Cyclosporine is a calcineurin phosphatase pathway inhibitor, used as an immunosuppressant drug to prevent rejection in organ transplantation. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1341
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.