Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva CyDye™ DIGE Fluor Minimal Dye Labeling Kits

Catalog No. 45002256 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
2 nmol of each: Cy2, Cy3, and Cy5
5 nmol of each: Cy2, Cy3, and Cy5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-256 5 nmol of each: Cy2, Cy3, and Cy5
45-002-245 2 nmol of each: Cy2, Cy3, and Cy5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-002-256 Supplier Cytiva Supplier No. 25801065
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Allow detection of up to three prelabeled protein samples and standards on the same 2-D electrophoresis gel. Amersham CyDye™ DIGE Fluor Minimal Labeling Kits contain size- and charge-matched fluorescent dyes specifically designed for detecting protein abundance differences in 2-D Fluorescence Difference Gel Electrophoresis using Ettan DIGE system.

  • Size- and charge-matched dyes enable co-migration of labeled samples within the gel
  • Bright and highly sensitive dyes allow the use of the minimal labeling technique
  • Minimal loss of signal during labeling, separation, and scanning
  • No change in signal over wide pH range used during first-dimension (IEF) separation
  • Discrete signal from each fluor with minimal cross-talk contributes to high accuracy

All minimal dyes allow the labeling of 50μg protein per labeling reaction. Minimal Dye Labeling Kits contain 2nmol or 5nmol of each of the minimal dyes.

For general 2-D applications where sufficient amounts of sample are available.

TRUSTED_SUSTAINABILITY

Specifications

Format Solid
For Use With (Application) For general 2-D applications where sufficient amounts of sample are available, detection of up to three prelabeled protein samples and standards on the same 2D electrophoresis gel
Quantity 5 nmol of each: Cy2, Cy3, and Cy5
Product Type Amersham CyDye DIGE Fluor Minimal Labeling Kit
Kit Contents 5nM of CyDye DIGE Fluor minimal labeling kit (Cy2, Cy3 and Cy5)
Content And Storage -15°C to -30°C
Color Blue, Green, Red
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.