Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Cylindrical Cow Magnet
SDPSCI_ED

Catalog No. S06980 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S06980 5+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S06980 Supplier United Scientific™ Supplier No. MGTCOW
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cylindrical Cow Magnet for magnetic field demonstrations is the classroom.

  • Cylindrical shaped magnet fed to cows to attract metal particles they may have ingested while grazing.
  • Use for a variety of investigations.

Order Info

L x dia.: 3 x 0.8 in. (7.5 x 2.0 cm); Weight: 0.22 lb. (100 g)

Specifications

Grade 5+
Format Set
Product Type Cow Magnet
Diameter (English) Ring 0.8 in.
Languages English
For Use With (Application) Use it in the classroom for magnetic field demonstration and other interesting experiments
Subject Physics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.