Learn More
TARGETMOL CHEMICALS INC CYM50260 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T15031
Encompass_Preferred
Also available in 1mg 10mg 25mg 50mg 100mg and bulk. Please contact Fisher for quotes. CYM50260 is a potent and exquisitely selective agonist of sphingosine-1-phosphate 4 receptor (S1P4-R EC50= nM) with no activity against S1P1-R S1P2-R S1P3-R and S1P5-R. purity: 99%
Catalog No. 50-257-7367
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.