Learn More
R&D Systems™ Cyno Monkey CD25/IL-2R alpha(NP_001270633.1) VersaClone cDNA
Shop All Novus Biologicals Products

Specifications
Specifications
| Gene | CD25/IL-2 R alpha |
| Accession Number | NP_001270633.1 |
| Gene ID (Entrez) | 3559 |
| Gene Alias | CD25, CD25 antigen, IDDM10, IL-2 R alpha, IL-2 receptor subunit alpha, IL2R, IL2R alpha, IL-2 R subunit alpha, IL-2 Ra, IL2RA, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Cynomolgus Monkey |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.