Learn More
Specifications
Specifications
| Gene | Common beta Chain |
| Accession Number | XP_005567452.1 |
| Gene ID (Entrez) | 1439 |
| Gene Alias | beta, low-affinity, CD131, CD131 antigen, CDw131, colony stimulating factor 2 receptor, beta, low-affinity(granulocyte-macrophage), CSF2RB, cytokine receptor common subunit beta, GM-CSF R beta, GM-CSF/IL-3/IL-5 receptor common beta subunit, GM-CSF/IL-3/IL-5 receptor common beta-chain, IL-3 R beta, IL3RB, IL-5 R beta, interleukin 3 receptor/granulocyte-macrophage colony stimulating factor 3 receptor, beta (high affinity) |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Cynomolgus Monkey |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
