Learn More
R&D Systems™ Cyno Monkey Neprilysin/CD10 (XP_005546216.1) VersaClone cDNA
Shop All Novus Biologicals Products

Specifications
Specifications
| Gene | Neprilysin/CD10 |
| Accession Number | XP_005546216.1 |
| Gene ID (Entrez) | 4311 |
| Gene Alias | Atriopeptidase, CALLA, CALLAmembrane metallo-endopeptidase (neutral endopeptidase, enkephalinase), CD10, CD10 antigen, CD10), CD10membrane metallo-endopeptidase variant 1, Common acute lymphocytic leukemia antigen, DKFZp686O16152, EC 3.4.24, EC 3.4.24.11, Enkephalinase, EPN, Leu-19, membrane metallo-endopeptidase, membrane metallo-endopeptidase variant 2, MGC126681, MGC126707, MME, NEPmembrane metallo-endopeptidase (neutral endopeptidase, enkephalinase, CALLA, neprilysin, Neutral endopeptidase, Neutral Endopeptidase 24.11, NKH1, SFE, Skin fibroblast elastase |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Cynomolgus Monkey |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.