Learn More
Description
- Signal regulatory protein gamma (SIRP gamma, designated CD172g), also called SIRP beta 2, is a type I transmembrane protein belonging to the SIRP/SHPS (CD172) family within the Ig superfamily.
- SIRP gamma is expressed on T cells, CD56bright NK cells and activated NK cells, but not myeloid cells.
- Like SIRP alpha, it binds CD47, but unlike SIRP alpha, it does not contain an obvious signaling mechanism.
- T cell SIRP gamma may play a role as an accessory protein interacting with CD47-expressing antigen presenting cells, and appears to promote antigen-specific T cell proliferation and costimulate T cell activation.
- The SIRP gamma/CD172g insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | SIRP gamma/CD172g |
| Accession Number | XP_005568591 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1177 bp |
| Gene ID (Entrez) | 55423 |
| Gene Alias | bA77C3.1, CD172g, CD172g antigen, FLJ42230, signal-regulatory protein beta 2, Signal-regulatory protein beta-2, signal-regulatory protein gamma, SIRP beta 2, SIRP-b2, SIRPB2CD172 antigen-like family member B, SIRP-beta-2, SIRPG, SIRPgamma, SIRP-gamma |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Cynomolgus Monkey |
| Product Line | R&D Systems |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
