Learn More
Bon Opus Biosciences Recombinant Cynomolgus ROR1 (C-hFc) -50ug
SDP
Supplier:  Bon Opus Biosciences BP22250UG
Encompass_Preferred
Recombinant Cynomolgus ROR1 is produced by our Mammalian expression system and the target gene encoding Ser25-Tyr392 is expressed with a Fc tag at the C-terminus. Expression system: Human 293 Cells. Accession# A0A2K5WTX7. Formulation: Supplied as a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# BP222
Catalog No. 50-208-0307
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.