Learn More
Bon Opus Biosciences Recombinant Cynomolgus OX40L/TNFSF4 (N-His) -50ug
SDP
Supplier:  Bon Opus Biosciences BP15450UG
Encompass_Preferred
Recombinant Cynomolgus OX40 Ligand is produced by our Mammalian expression system and the target gene encoding Gln51-Leu183 is expressed with a 8His tag at the N-terminus. Expression system: Human 293 Cells. Accession# F7FL80. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Bon Opus Cat# BP154
Catalog No. 50-208-0243
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.