Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Cynomolgous Monkey Hepassocin (EHH64014.1) VersaClone cDNA
SDP

Catalog No. RDC3087 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDC3087 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RDC3087 Supplier R&D Systems™ Supplier No. RDC3087
Add to Cart
Add to Cart
This item is not returnable. View return policy

Contains gene inserts that are fully sequenced and produced with high quality

Specifications

Gene Hepassocin/FGL1
Accession Number EHH64014.1
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Insert Type 952 bp
Gene ID (Entrez) 2267
Gene Alias FGL1, fibrinogen-like 1, fibrinogen-like protein 1, Fibrinogen-like-protein 1, FREP1, Hepassocin, hepatocellular carcinoma-related sequence, Hepatocyte-derived fibrinogen-related protein 1, HFREP-1, HFREP1MGC12455, HP-041, LFIRE1, LFIRE-1, Liver fibrinogen-related protein 1
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Cynomolgus Monkey
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.