Learn More
ACROBiosystems ACROBiosystems Cynomolgus 4-1BB Ligand / TNFSF9 Protein, Fc Tag
Supplier:  ACROBiosystems 41LC5254100UG
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus 4-1BB Ligand, Fc Tag (41L-C5254) is expressed from human 293 cells (HEK293). It contains AA Leu 67 - Glu 251 (Accession # XP_015296398.1).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 87.6 kDa.
Catalog No. 50-210-6420