Learn More
ACROBiosystems Cynomolgus ASGR1 1mg
Supplier:  ACROBiosystems AS1C52431MG
Encompass_Preferred
Cynomolgus ASGR1 Protein His Tag (AS1-C5243) is expressed from human 293 cells (HEK293). It contains AA Ser 61 - Leu 291 (Accession # XP_005582755.1).
Catalog No. 50-253-5490
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.