Learn More
Sino Biological Cynomolgus CD3D Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90046C02H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus CD3D (Q95LI8) (Met1-Ala105) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-2307