Learn More
Sino Biological Cynomolgus Claudin-6 Protein (VLP, Full-Length, Flag Tag) 20µg
Supplier:  Sino Biological 91074C25H20
Encompass_Preferred
A DNA sequence encoding the Cynomolgus Claudin-6-VLP (Full Length) (G7Q0B0) (Met1-Val220) was fused with a signal peptide at the N-terminus and a flag tag at the C-terminus. This protein was prepared using our VLP platform.As an innovative technology platform, VLPs are generated through the self-assembly of viral proteins in HEK293 cells. This process efficiently integrates membrane proteins onto the VLP surface, preserving their native conformation and yielding soluble, high-concentration proteins that are ideally suited for immunization and antibody screening.
Catalog No. 50-000-83632
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.