Learn More
ACROBiosystems Cynomolgus IL-12 R beta 1 1mg
Supplier:  ACROBiosystems ILBC52531MG
Encompass_Preferred
Cynomolgus IL-12 R beta 1 Protein Fc Tag (ILB-C5253) is expressed from human 293 cells (HEK293). It contains AA Cys 24 - Asp 545 (Accession # G7PWY9-1).
Catalog No. 50-253-4588
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.