Learn More
KACTUSBIO INC Cynomolgus IL-18BP Protein (IL8-CM1BP) 100UG
Supplier:  KACTUSBIO INC IL8CM1BP100UG
Encompass
Recombinant Cynomolgus IL-18BP Protein is expressed from HEK293 with His tag at the C-Terminus.It contains Thr28-Pro207.
Catalog No. NC2953548