Learn More
Sino Biological Cynomolgus IL13 PAb 100UL 11057-RP02-100
Supplier:  Sino Biological 11057RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Cynomolgus IL-13 (rC IL-13; Catalog#11057-C07H; ABG75889.1; Ser 21-Asn 132). IL-13 specific IgG was purified by Cynomolgus IL-13 affinity chromatography.
Catalog No. 50-240-8518
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.