Learn More
ACROBiosystems ACROBiosystems Cynomolgus LILRB4 / CD85k / ILT3 Protein, His Tag
Supplier:  ACROBiosystems CDKC5227100UG
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus LILRB4, His Tag (CDK-C5227) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Glu 259 (Accession # XP_015297198.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.0 kDa. The protein migrates as 34-42 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus LILRB4, His Tag (Cat. No. CDK-C5227) at 2 μg/mL (100 μL/well) can bind Human ANGPTL7, His Tag (Cat. No. AN7-H52H5) with a linear range of 8-125 ng/mL (QC tested).
Catalog No. 50-210-6586
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.