Learn More
R&D Systems™ Cynomolgus Monkey 20S Immunoproteasome Protein, Carrier-free
Description
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | Xmg/mL (X μM) in 50mM HEPES pH 7.6, 100mM NaCl, 1mM DTT |
| Molecular Weight (g/mol) | 700 kDa |
| Quantity | 25μg |
| Source | Cynomolgus monkey spleen-derived 20S Immunoproteasome protein |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Regulatory Status | RUO |
| Common Name | 20S Immunoproteasome |
| Biological Activity | 20S Proteasome is able to degrade substrates in an ATP-independent manner. It can be activated chemically with SDS (0.035%, final) or by the addition of PA28. Reaction conditions will need to be optimized for each specific application. Initial 20S Prote |
| Conjugate | Unconjugated |
| Show More |
For Research Use Only. Not for use in Humans.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.