Learn More
R&D Systems™ Cynomolgus Monkey CCL4/MIP-1 beta (Unique) VersaClone cDNA
Shop All Novus Biologicals Products

Description
- CCL4 is a member of the CC or beta chemokine subfamily.
- CCL4 is expressed primarily by T cells, B cells, and monocytes after antigen or mitogen stimulation.
- The functional receptor for CCL4 has been identified as CCR5.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | cynoLOC102117861 |
| Accession Number | unique |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 292 bp |
| Gene ID (Entrez) | 6351 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Cynomolgus Monkey |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.