Learn More
R&D Systems™ Cynomolgus Monkey CD43 (EHH60309.1) VersaClone cDNA
Shop All Novus Biologicals Products

Specifications
Specifications
| Gene | CD43/Sialophorin |
| Accession Number | EHH60309.1 |
| Gene ID (Entrez) | 6693 |
| Gene Alias | CD43 antigen, CD43), Galactoglycoprotein, GALGP, Leukocyte sialoglycoprotein, Sialophorin, sialophorin (gpL115, leukosialin, CD43) |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.