Learn More
R&D Systems™ Cynomolgus Monkey CEACAM-5/CD66e (unique) VersaClone cDNA
Shop All Novus Biologicals Products

Specifications
Specifications
| Gene | CEACAM5/CD66e |
| Accession Number | unique |
| Gene ID (Entrez) | 1048 |
| Gene Alias | Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e, CD66e antigen, CEA, CEACAM5, CEACD66e, DKFZp781M2392, Meconium antigen 100 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Cynomolgus Monkey |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.