Learn More
Description
- Also known as: IL-12/IL-35 p35; IL-12A; IL35p35
- IL-12 p35 is a subunit of a cytokine that acts on T and natural killer cells and has a broad array of biological activities.
- This cytokine is required for the T-cell-independent induction of interferon (IFN)-gamma and is important for the differentiation of both Th1 and Th2 cells.
- The responses of lymphocytes to this cytokine are mediated by the activator of transcription protein STAT4.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | cynoIL12A |
| Accession Number | unique |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 673 bp |
| Gene ID (Entrez) | 3592 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Cynomolgus Monkey |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
