Learn More
Description
- ITGA2B integrin subunit alpha 2b [ Macaca fascicularis (crabeating macaque) ]
- The Integrin alpha 2b/CD41 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- Integrins are heterodimeric integral cell-surface proteins composed of an alpha and a beta chain known to participate in cell adhesion as well as cell-surface mediated signaling.
- ITGA2B undergoes posttranslational cleavage to yield disulfide-linked light and heavy chains that join with beta-3 to form a fibronectin receptor expressed in platelets that play a crucial role in coagulation.
- Mutations that interfere with this role may result in Glanzmann thrombasthenia.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | cynoITGA2B |
| Accession Number | unique |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 3133 bp |
| Gene ID (Entrez) | 3674 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Cynomolgus Monkey |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
