Learn More
Description
- ITGAV integrin subunit alpha V [ Macaca fascicularis (crabeating macaque) ]
- Integrins are heterodimeric integral cell-surface proteins known to participate in cell adhesion as well as cellsurface mediated signaling, mediating interactions between adhesion molecules on adjacent cells and/or the extracellular matrix (ECM)
- Although they have no catalytic activity, integrins can be part of multimolecular signaling complexes known as focal adhesions
- ITGAV undergoes posttranslational cleavage to yield disulfide-linked heavy and light chains that combine with multiple integrin beta chains to form different integrins
- ITGAV has been shown to heterodimerize with beta 1, beta 3, beta 5, beta 6, and beta 8
- It is proposed that ITGAV may regulate angiogenesis and cancer progression
- Integrins have diverse roles in several biological processes including cell migration during development and wound healing, cell differentiation, and apoptosis. Their activities can also regulate the metastatic and invasive potential of tumor cells
- The Integrin alpha V/CD51 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | cynoITGAV |
| Accession Number | XP_005573729 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 3160 bp |
| Gene ID (Entrez) | 3685 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Cynomolgus Monkey |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
