Learn More
Description
- Integrins are integral cellsurface proteins composed of an alpha chain and a beta chain.
- They are known to participate in cell adhesion as well as cellsurface mediated signaling.
- ITGB2 belongs to the integrin beta chain family of proteins.
- It combines with the alpha L chain to form the integrin LFA-1 and combines with the alpha M chain to form the integrin Mac1.
- Defects in ITGB2 are the cause of leukocyte adhesion deficiency type I (LAD1).
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | cynoITGB2 |
| Accession Number | XP_005548577 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 2323 bp |
| Gene ID (Entrez) | 3689 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Cynomolgus Monkey |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
