Learn More
Description
- ITGB3 integrin subunit beta 3 [ Macaca fascicularis (crabeating macaque) ]
- The Integrin beta 3/CD61 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- Integrins are integral cell-surface proteins known to participate in cell adhesion as well as cell-surface mediated signaling.
- They are composed of an alpha chain and a beta chain.
- ITGB3 is the integrin beta chain that forms a heterodimer with alpha-IIb or alpha-V chains.
- It is involved in platelet aggregation, which can repair damaged endothelial cells, such as those found in Glanzmann thrombasthenia.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | cynoITGB3 |
| Accession Number | XP_005584667 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 2380 bp |
| Gene ID (Entrez) | 3690 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Cynomolgus Monkey |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
