Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Cynomolgus Monkey JAM-A (NP_001270452) VersaClone cDNA
SDP

Catalog No. RDC2748 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDC2748 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RDC2748 Supplier R&D Systems™ Supplier No. RDC2748

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.

  • JAM-A belongs to the immunoglobulin superfamily and the junctional adhesion molecular (JAM) family.
  • JAM-A is a transmembrane receptor that is predominantly expressed at intercellular junctions of both epithelial and endothelial cells.
  • JAMA exhibits homophilic interactions to regulate tight junction assembly and modulate paracellular permeability.
  • This homophilic interaction also mediates platelet aggregation and adhesion to endothelial cells and may play a role in thrombosis.
  • JAMA is also a ligand for the integrin LFA1, involved in leukocyte transmigration, and a receptor for reovirus.
  • The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
  • For reasearch use only. Not for use in humans.

Specifications

Gene cynoF11R
Accession Number NP_001270452
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Insert Type 913 bp
Gene ID (Entrez) 50848
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Cynomolgus Monkey
Product Line R&D Systems
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.