Learn More
R&D Systems™ Cynomolgus Monkey Lgr5/GPR49 (XP_005571599) VersaClone cDNA
Shop All Novus Biologicals Products

Description
- LGR5 leucine rich repeat containing G protein-coupled receptor 5 [ Macaca fascicularis (crab-eating macaque) ]
- The Lgr5/GPR49 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- LGR5 is a member of the G protein-coupled receptor 1 family and is the receptor for R-spondins that mediate the potentiation of Wnt/β-catenin signaling by enhancing Wntinduced LRP6 phosphorylation.
- This receptor is expressed in the gonads, the adrenal gland, and in the brain.
- LGR5 acts as a stem cell marker of the intestinal epithelium and the hair follicle.
- Increased expression of LGR5 has been linked to ovarian, colon and hepatocellular cancers.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | cynoLGR5 |
| Accession Number | XP_005571599 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 2737 bp |
| Gene ID (Entrez) | 8549 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Cynomolgus Monkey |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.