Learn More
ACROBiosystems Cynomolgus Mucin-1 / MUC-1 Protein, Fc Tag
Supplier:  ACROBiosystems MU1C5252100UG
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus Mucin-1, Fc Tag (MU1-C5252) is expressed from human 293 cells (HEK293). It contains AA Leu 254 - Gly 373 (Accession # XP_005541632.1).;This protein carries a human IgG1 Fc tag at the C-terminus.The mature Mucin-1 is a non-covalent heterodimeric complex with the proteolytically cleaved N-terminal fragment and C-terminal fragment. Each N-terminal fragment and C-terminal fragment has a calculated MW of 7.2 kDa and 32.8 kDa. The protein migrates as 48 kDa (precursor protein) and 38-43 kDa (C-terminus) under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-205-4943
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.