Learn More
ACROBiosystems ACROBiosystems Cynomolgus NKp46 / NCR1 / CD335 Protein, His Tag (MALS verified)
Supplier:  ACROBiosystems NC1C52H4100UG
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus NKp46, His Tag (NC1-C52H4) is expressed from human 293 cells (HEK293). It contains AA Pro 22 - Arg 257 (Accession # Q95JB9-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.6 kDa. The protein migrates as 34-40 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6531
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.