Learn More
ACROBiosystems ACROBiosystems Cynomolgus PD-1 / PDCD1 Protein, Fc Tag
Supplier:  ACROBiosystems PD1C52541MG
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus PD-1, Fc Tag (PD1-C5254) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # B0LAJ3).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.6 kDa. The protein migrates as 53-60 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.;Immobilized Human PD-L1, Mouse IgG1 Fc Tag, low endotoxin (HPLC-verified) (Cat. No. PD1-H52A3) at 5 μg/mL (100 μL/well) can bind Cynomolgus PD-1, Fc Tag (Cat. No. PD1-C5254) with a linear range of 3-33 ng/mL (QC tested).;Immobilized Cynomolgus PD-1, Fc Tag (Cat. No. PD1-C5254) at 2 μg/mL (100 μL/well) can bind Nivolumab with a linear range of 0.1-3 ng/mL (Routinely tested).
Catalog No. 50-210-6740
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.