Learn More
Sino Biological Recombinant Cynomolgus, Rhesus Erythropoietin R/EPOR Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 90962C08H100
Encompass_Preferred
A DNA sequence encoding the Cynomologus / Rhesus EPOR (EHH59220.1 & XP_001105833.2) (Met1-Pro250) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-299-2729