Learn More
ACROBiosystems ACROBiosystems Cynomolgus / Rhesus macaque Cadherin-6 / KCAD / CDH6 Protein, His Tag
Supplier:  ACROBiosystems CA6C52H3100UG
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus / Rhesus macaque Cadherin-6, His Tag (CA6-C52H3) is expressed from human 293 cells (HEK293). It contains AA Thr 22 - Gly 614 (Accession # G7MUQ7-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 67.9 kDa. The protein migrates as 80-95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6252
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.