Learn More
ACROBiosystems ACROBiosystems Cynomolgus / Rhesus macaque TPBG / 5T4 Protein, His Tag
Supplier:  ACROBiosystems TPGC52H31MG
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus / Rhesus macaque TPBG, His Tag (TPG-C52H3) is expressed from human 293 cells (HEK293). It contains AA Thr 35 - Ser 355 (Accession # Q4R8Y9-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.7 kDa.
Catalog No. 50-210-6896
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.